Order Entry
Northern Ireland
ContactUsLinkComponent
 
Pesticide reference standard, Pyribenzoxim, neat, Honeywell Fluka™
Catalog # HONCR1122-50MG
CAS Number:  
Pesticide reference standard, Pyribenzoxim, neat, Honeywell Fluka™
Catalog # HONCR1122-50MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mg
  • CAS No.:
    168088-61-7
  • Description:
    Pyribenzoxim
  • Formula:
    C₃₂H₂₇N₅O₈
  • Storage temperature:
    Ambient
  • Description:
    Pyribenzoxim
  • Freezing point:
    119 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results