Pesticide reference standard, Tecloftalam imide, neat, Honeywell Fluka™
Catalog # HONCR1037-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:26491-30-5
- Description:Tecloftalam imide
- Melting point:252...256 °C
- Formula:C₁₄H₃Cl₆NO₂
- Storage temperature:2...8 °C
- Description:Tecloftalam imide
- Freezing point:252...256 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results