Order Entry
Northern Ireland
ContactUsLinkComponent
Sodium carbonate monohydrate ≥99.5%, granular, AR® ACS, Macron Fine Chemicals™
Irritant
Sodium carbonate monohydrate ≥99.5%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # MACR7516-04
CAS Number:  
undefined
Irritant
Sodium carbonate monohydrate ≥99.5%, granular, AR® ACS, Macron Fine Chemicals™
Catalog # MACR7516-04
CAS Number:  

Specifications

  • Pack type:
    Poly/plastic bottle, Polystormor®
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • ACS - Assay (Na₂CO₃·H₂O) (by acidimetry):
    ≥99.5 %
  • ACS - Loss on Drying at 150°C:
    13.0 - 15.0 %
  • ACS - Insoluble Matter:
    ≤0.01 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Phosphate (PO₄):
    ≤5 ppm
  • ACS - Silica (SiO₂):
    ≤0.005 %
  • ACS - Sulfur Compounds (as SO₄):
    ≤0.004 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - Calcium (Ca):
    ≤0.03 %
  • ACS - Magnesium (Mg):
    ≤0.005 %
  • ACS - Potassium (K):
    ≤0.005 %
  • ACS - Solubility (8 in 50):
    Passes Test

Specification Test Results

About this item

 :
 :124.00 g/mol
Melting Pt:107,85 °C
Density:2,25 g/cm³ (20 °C)
EINECS:207-838-8
 :GHS07
Storage Temperature:Ambient
CAS Number:5968-11-6
Formula:Na₂CO₃·1H₂O
MDL Number:MFCD00149177