
Molecular sieve dehydrate, beads 10-20 mesh for drying solvents, with indicator, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
:
MDL Number:MFCD00147630
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Particle Size:10-20 mesh
- Appearance (color):complying
- Loss on ignition (900°C):Max. 3 %
- Grain size 1-3 mm:Min. 95 %
Specification Test Results
Frequently Bought Together