Diatomaceous earth, powder suitable for most filtrations, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: Diatomaceous silica, Kieselguhr, Infusorial earth, Diatomite, Celite
CAS Number:68855-54-9
Formula:SiO₂
MDL Number:MFCD00132803
Density:2.4 g/cm³
Molecular Weight:60.08 g/mol
Melting Pt:450 °C
EINECS:293-303-4
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance of the substance:complying
Specification Test Results
Frequently Bought Together