Order Entry
Northern Ireland
ContactUsLinkComponent
 
EDTA disodium salt solution with zinc complex added (solution B) concentrate for dilution to 1L for hardness of water determinations, for complexometry (1 ml = 1°dH/100 ml H₂O), Fluka™
Catalog # 38056-1EA
CAS Number:  
EDTA disodium salt solution with zinc complex added (solution B) concentrate for dilution to 1L for hardness of water determinations, for complexometry (1 ml = 1°dH/100 ml H₂O), Fluka™
Catalog # 38056-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
Boiling Pt:100 °C
Storage Temperature:Ambient
MDL Number:MFCD00147223