Order Entry
Northern Ireland
ContactUsLinkComponent
 
Test strips, ammonia
Catalog # MANA90714
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, ammonia
Catalog # MANA90714
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Ammonium and ammonia test strips
  • Tests per kit:
    25
  • Range:
    0 - 6 mg/l NH₄+

Specifications

About this item

These test strips are for the fast and reliable determination of Ammonium, respectively Ammonia, in solutions. The easy dip-and-read procedure provides safe results within 40 seconds. Ammonium/Ammonia is harmful to fish and other aquatic life in aquariums. This test shows the amount of Ammonium quickly and easily.

  • The aquarium can be kept in good condition and fish stay healthy
  • Results within seconds: Just dip, wait and read against multi-stage colour scale
  • Always ready to use: No accessories necessary for evaluation
  • Safe results: Desiccant in the stopper for optimal protection of the strips against humidity

Always ready to use, no accessories necessary for evaluation. Safe results since the desiccant in the stopper for optimal protection of the strips against humidity.

Colour reaction: Bright yellow to blue