Multicomponent alcohol calibration kit (C1-C3) solution, Cerilliant®, Supelco®
Catalog # A-054-3X1.2ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:varied, in water
- Pk:3
- Description:Multicomponent alcohol calibration kit (C1-C3) solution
- Pk Info:3 ampoules/kit
Specifications
About this item
Multi-component calibration kits containing acetone, ethanol, isopropanol, and methanol varying concentrations. These kits ares suitable for calibration of blood alcohol testing methods using GC Headspace.
- SNAP-N-SPIKE®/SNAP-N-SHOOT®
Each component at stated concentration (μg/mL),
C1 C2 C3
Acetone 500 1000 4000
Ethanol 500 1000 4000
Isopropanol 500 1000 4000
Methanol 500 1000 4000
Caution: Storage temperature: 2 to 8 °C (refrigerator)