Multi-component alcohol mixture for calibration, Cerilliant®, Supelco®
Catalog # A-127-6X1.2ML
Supplier: Merck
CAS Number:
Specifications
- Pk:6
- Description:Multi-component alcohol mix
Specifications
About this item
A multicomponent calibration kit containing acetone, ethanol, isopropanol, and methanol at concentrations of 100, 250, 500, 1000, 2000, and 4000 μg/mL. This kit is suitable for calibration of blood alcohol testing methods using GC Headspace.
- SNAP-N-SPIKE®/SNAP-N-SHOOT®
Caution: Storage temperature: 2 to 8 °C (refrigerator)