

Iodide/Iodate concentrate for dilution to 1L, 0.05 M I2 (0.1N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
:
MDL Number:MFCD00011355
Density:1,160 g/cm³ (20 °C)
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
Frequently Bought Together