Order Entry
Northern Ireland
ContactUsLinkComponent
DMNP-EDTA tetrapotassium salt (Caged Calcium) ≥90% (by HPLC)
DMNP-EDTA tetrapotassium salt (Caged Calcium) ≥90% (by HPLC)
Catalog # 50050.
Supplier:  Biotium
CAS Number:  
DMNP-EDTA tetrapotassium salt (Caged Calcium) ≥90% (by HPLC)
Catalog # 50050.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :626 g/mol
Formula:C₁₈H₁₉K₄N₃O₁₂