N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine ≥95% (by TLC) ion chelator
Catalog # 59003.
Supplier: Biotium
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:424.55 g/mol
Storage Temperature:Refrigerator
CAS Number:16858-02-9
Formula:C₂₆H₂₈N₆
MDL Number:MFCD00036918