Order Entry
Northern Ireland
ContactUsLinkComponent
BAPTA tetracesium salt ≥95% (by TLC) ion chelator, for calcium determination
Irritant
BAPTA tetracesium salt ≥95% (by TLC) ion chelator, for calcium determination
Catalog # 50001.
Supplier:  Biotium
CAS Number:  
Irritant
BAPTA tetracesium salt ≥95% (by TLC) ion chelator, for calcium determination
Catalog # 50001.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :1004 g/mol
 :GHS07
CAS Number:480436-84-8
Formula:C₂₃H₂₄Cs₄N₂O₁₀