Supelco® TraceCERT® Organic Reference Standard, EPA 8270 Semivolatile Internal Standard Mix
Supplier: Merck
Certificates
About this item
- GHS signal: GHS08, GHS07, GHS09
Caution: P202 - P261 - P273 - P302 + P352 - P305 + P351 + P338 - P308 + P313
Specifications
- Storage conditions:2…8 °C
- Hazard class description:H315 - H319 - H336 - H350 - H411
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
Specifications
Frequently Bought Together