Zinquin free acid ≥98% (by HPLC) fluorescent indicator, for the determination of zinc
Catalog # 52022.
Supplier: Biotium
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:386.42 g/mol
Storage Temperature:Refrigerator
CAS Number:151606-29-0
Formula:C₁₉H₁₈N₂O₅S
MDL Number:MFCD03452711