Order Entry
Northern Ireland
ContactUsLinkComponent
Zinquin ethyl ester ≥95% (by HPLC) fluorescent indicator, for the determination of zinc
Zinquin ethyl ester ≥95% (by HPLC) fluorescent indicator, for the determination of zinc
Catalog # 52020.
Supplier:  Biotium
CAS Number:  
undefined
Zinquin ethyl ester ≥95% (by HPLC) fluorescent indicator, for the determination of zinc
Catalog # 52020.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :414.47 g/mol
Storage Temperature:Refrigerator
CAS Number:181530-09-6
Formula:C₂₁H₂₂N₂O₅S