Order Entry
Northern Ireland
ContactUsLinkComponent
 
Irritant
6-Carboxyfluorescein ≥98% (by HPLC) fluorescent pH indicator
Catalog # 51020.
Supplier:  Biotium
CAS Number:  
undefined
Irritant
6-Carboxyfluorescein ≥98% (by HPLC) fluorescent pH indicator
Catalog # 51020.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :376.32 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:3301-79-9
Formula:C₂₁H₁₂O₇
MDL Number:MFCD00036873