Order Entry
Northern Ireland
ContactUsLinkComponent
5-Carboxyfluorescein ≥98% (by HPLC) fluorescent pH indicator
Irritant
5-Carboxyfluorescein ≥98% (by HPLC) fluorescent pH indicator
Catalog # 51019.
Supplier:  Biotium
CAS Number:  
undefined
Irritant
5-Carboxyfluorescein ≥98% (by HPLC) fluorescent pH indicator
Catalog # 51019.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :376.32 g/mol (anhy)
 :GHS07
Storage Temperature:Refrigerator
CAS Number:76823-03-5
Formula:C₂₁H₁₂O₇.xH₂O
MDL Number:MFCD00036874