Order Entry
Northern Ireland
ContactUsLinkComponent
5(6)-Carboxyfluorescein ≥95% (by HPLC) fluorescent pH indicator
5(6)-Carboxyfluorescein ≥95% (by HPLC) fluorescent pH indicator
Catalog # 51013.
Supplier:  Biotium
CAS Number:  
undefined
5(6)-Carboxyfluorescein ≥95% (by HPLC) fluorescent pH indicator
Catalog # 51013.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :752.63 g/mol
CAS Number:72088-94-9
Formula:C₄₂H₂₄O₁₄
MDL Number:MFCD00151081