Order Entry
Northern Ireland
ContactUsLinkComponent
 
Phytochemical standard, Valproic Acid-D6 solution, (CRM), Supelco®
  V-029-1ML
 :  Merck
 :  
undefined
Phytochemical standard, Valproic Acid-D6 solution, (CRM), Supelco®
  V-029-1ML
 :  Merck
 :  

 

  • Concentration:
    1 mg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    87745-18-4
  • Description:
    Valproic Acid-D6 solution
  • Description:
    Valproic Acid-D6 solution
  • Pk Info:
    Ampoule of 1 ml