Explosives standard, 2-Mononitroglycerin solutions, (CRM), Supelco®
M-078-1ML
: Merck
:
- Concentration:1,0 mg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:620-12-2
- Description:2-Mononitroglycerin solution
- Description:2-Mononitroglycerin solution
- Pk Info:Ampoule of 1 ml