Explosives standard, 1-Mononitroglycerin solutions, (CRM), Supelco®
M-077-1ML
: Merck
:
- Concentration:1,0 mg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:624-43-1
- Description:1-Mononitroglycerin solution
- Description:1-Mononitroglycerin solution
- Pk Info:Ampoule of 1 ml