Supelco® Cerilliant® Organic Reference Standard, Tetramethylenedisulfotetramine solution
Catalog # ERT-113S-1.2ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:1,2 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:80-12-6
- Description:Tetramethylenedisulfotetramine solution
- Storage conditions:–20 °C
- Hazard class description:H225 - H302 + H312 + H332 - H319
- Shipping conditions:STD
- MDL No.:MFCD00221771
- M-Clarity quality level:MQ300
- Molecular weight:240,26
- Flash point:2 °C - Closed cup
- Pk Info:Ampule of 1,2 ml
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338