Explosives standard, Pentaerythritol tetranitrate solution, (CRM), Supelco®
Catalog # ERP-109S-5ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:10 mg/ml in acetonitrile
- Pk:5 mL
- Standards:Certified reference material
- CAS No.:78-11-5
- Description:Pentaerythritol tetranitrate solution
- Description:Pentaerythritol tetranitrate solution
- Pk Info:Ampoule of 5 ml