Order Entry
Northern Ireland
ContactUsLinkComponent
 
Explosives standard, 1,3-Dinitroglycerin solution, (CRM), Supelco®
Catalog # D-003-1ML
Supplier:  Merck
CAS Number:  
undefined
Explosives standard, 1,3-Dinitroglycerin solution, (CRM), Supelco®
Catalog # D-003-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    623-87-0
  • Description:
    1,3-Dinitroglycerin solution
  • Description:
    1,3-Dinitroglycerin solution
  • Pk Info:
    Ampoule of 1 ml

Specifications

About this item