Supelco® Organic Reference Standard, Ethylene oxide solution
Catalog # CRM47949
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 µg/ml in dichloromethane
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:75-21-8
- Description:Ethylene oxide solution
- Environmentally Preferable:
- Storage conditions:2…8 °C
- Hazard class description:H315 - H319 - H336 - H340 - H350
- Shipping conditions:WICE
- MDL No.:MFCD00014482
- M-Clarity quality level:MQ100
- Synonym(s):Ethylene oxide solution
- Molecular weight:44,05
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS08, GHS07
Caution: P202 - P261 - P264 - P302 + P352 - P305 + P351 + P338 - P308 + P313