Organic reference standards, 2-Methylisoborneol solution, (CRM), TraceCERT®, Supelco®
Catalog # CRM47523
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:2371-42-8
- Description:2-Methylisoborneol solution
- Description:2-Methylisoborneol solution
- Pk Info:Ampoule of 1 ml