Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Benzaldehyde-DNPH solution
Supelco® Organic Reference Standard, Benzaldehyde-DNPH solution
Catalog # CRM47343
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Benzaldehyde-DNPH solution
Catalog # CRM47343
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in acetonitrile (as the aldehyde)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    1157-84-2
  • Description:
    Benzaldehyde-DNPH solution
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00156353
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Benzaldehyde-2,4-dinitrophenylhydrazone solution; Benzaldehyde-2,4-DNPH solution
  • Flash point:
    2,0 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338