Organic reference standards, Acrolein-DNPH solution, (CRM), Supelco®
Catalog # CRM47342
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 µg/ml in acetonitrile (as the aldehyde)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:888-54-0
- Description:Acrolein-DNPH solution
- Description:Acrolein-DNPH solution
- Pk Info:Ampoule of 1 ml