Order Entry
Northern Ireland
ContactUsLinkComponent
 
Organic reference standards, DBA Isocyanate Monomers Mix, (CRM), TraceCERT®, Supelco®
Catalog # CRM40569
Supplier:  Merck
CAS Number:  
Organic reference standards, DBA Isocyanate Monomers Mix, (CRM), TraceCERT®, Supelco®
Catalog # CRM40569
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    in acetone (varied concentration)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • Description:
    DBA Isocyanate Monomers Mix
  • Description:
    DBA Isocyanate Monomers Mix
  • Pk Info:
    Ampoule of 1 ml

Specifications

About this item