Order Entry
Northern Ireland
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Dopamine Hydrochloride
  43658-50MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Dopamine Hydrochloride
  43658-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    62-31-7
  • Description:
    Dopamine hydrochloride
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H317
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00012898
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    2-(3,4-Dihydroxyphenyl)ethylamine hydrochloride; 3,4-Dihydroxyphenethylamine hydrochloride; 3-Hydroxytyramine hydrochloride; 4-(2-Aminoethyl)-1,2-benzenediol hydrochloride
  • Melting point:
    248...250 °C (lit.)
  • Formula:
    (HO)₂C₆H₃CH₂CH₂NH₂·HCl
  • Molecular weight:
    189,64
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS07, GHS09
 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
 : P261 - P272 - P280 - P302 + P352 - P333 + P313 - P362 + P364