Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Drug and metabolite standards, Cyclosporin A solutions, (CRM)
Supelco® Drug and metabolite standards, Cyclosporin A solutions, (CRM)
Catalog # C-093-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Drug and metabolite standards, Cyclosporin A solutions, (CRM)
Catalog # C-093-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1,0 mg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    59865-13-3
  • Description:
    Cyclosporin A solution
  • Description:
    Cyclosporin A solution
  • Pk Info:
    Ampoule of 1 ml

Specifications

About this item