Supelco® Drug and metabolite standards, Ascomycin solution, (CRM)
A-094-1ML
: Merck
:
- Concentration:1,0 mg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:104987-12-4
- Description:Ascomycin solution
- Description:Ascomycin solution
- Pk Info:Ampoule of 1 ml