Drug and metabolite standards, alpha-Hydroxyetizolam solution, (CRM), Supelco®
Catalog # H-129-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:64546-10-7
- Description:alpha-Hydroxyetizolam solution
- Description:alpha-Hydroxyetizolam solution
- Pk Info:Ampoule of 1 ml
Specifications
About this item
A major urinary metabolite of the benzodiazepine etizolam. Etizolam, commonly sold under the trade name Etilaam, Etizola, or Depas, is a benzodiazepine analog used for the short-term treatment of insomnia, anxiety, or panic attacks.
- Snap-N-Spike®/Snap-N-Shoot®
This certified Snap-N-Spike®; solution standard is suitable for use in LC/MS or GC/MS benzodiazepine testing methods in clinical toxicology, urine drug testing, or forensic analysis.
Caution: Storage temperature: −20 °C (Freezer)