Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Drug and metabolite standards, 17beta-Estradiol-2,3,4-13C3 solution, (CRM)
Supelco® Drug and metabolite standards, 17beta-Estradiol-2,3,4-13C3 solution, (CRM)
Catalog # E-117-1ML
Supplier:  Merck
CAS Number:  
Supelco® Drug and metabolite standards, 17beta-Estradiol-2,3,4-13C3 solution, (CRM)
Catalog # E-117-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    1261254-48-1
  • Description:
    17beta-Estradiol-2
  • Description:
    17beta-Estradiol-2
  • Pk Info:
    Ampoule of 1 ml

Specifications

About this item