Order Entry
Northern Ireland
ContactUsLinkComponent
5-CR110-PEO₄-propionate SE amine-reactive fluorescent dye
5-CR110-PEO₄-propionate SE amine-reactive fluorescent dye
Catalog # 90081.
Supplier:  Biotium
CAS Number:  
undefined
5-CR110-PEO₄-propionate SE amine-reactive fluorescent dye
Catalog # 90081.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :718.71 g/mol
Storage Temperature:Freezer
Formula:C₃₆H₃₈N₄O₁₂