6-Carboxytetramethylrhodamine succinimidyl ester (6-TAMRA SE) ≥98% (by TLC) amine-reactive fluorescent dye
Catalog # 90035.
Supplier: Biotium
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:527.52 g/mol
Storage Temperature:Freezer
CAS Number:150810-69-8
Formula:C₂₉H₂₅N₃O₇