5-Carboxyrhodamine 6G succinimidyl ester ≥90% (by HPLC) amine-reactive fluorescent dye
Catalog # 90032.
Supplier: Biotium
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:555.6 g/mol
Storage Temperature:Freezer
Formula:C₃₁H₂₉N₃O₇