5(6)-Carboxyrhodamine 110-X succinimudyl ester ≥90% (by TLC) amine-reactive fluorescent dye
Catalog # 90003.
Supplier: Biotium
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:584.58 g/mol
Storage Temperature:Freezer
Formula:C₃₁H₂₈N₄O₈