5(6)-Carboxyrhodamine 110-X hydrochloride ≥95% (by TLC) amine-reactive fluorescent dye
Catalog # 90002.
Supplier: Biotium
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:524 g/mol
Storage Temperature:Refrigerator
Formula:C₂₈H₃₀ClN₃O₆