Order Entry
Northern Ireland
ContactUsLinkComponent
 
5(6)-Carboxyrhodamine 110 succinimidyl ester ≥85% (by HPLC) amine-reactive fluorescent dye
Catalog # 90001.
Supplier:  Biotium
CAS Number:  
undefined
5(6)-Carboxyrhodamine 110 succinimidyl ester ≥85% (by HPLC) amine-reactive fluorescent dye
Catalog # 90001.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :471.42 g/mol
Storage Temperature:Freezer
Formula:C₂₅H₁₇N₃O₇