Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Lidocaine hydrochloride
Supelco® Organic Reference Standard, Lidocaine hydrochloride
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Lidocaine hydrochloride
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    6108-05-0
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H301
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00150329
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    Lidocaine hydrochloride monohydrate; 2-Diethylamino-N-(2,6-dimethylphenyl)acetamide hydrochloride monohydrate; Lignocaine hydrochloride monohydrate; Xylocaine hydrochloride monohydrate
  • Molecular weight:
    288,81

Specifications

Related Information

Frequently Bought Together