Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, ʟ-Histidine
Supelco® TraceCERT® Organic Reference Standard, ʟ-Histidine
Catalog # 73767-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® TraceCERT® Organic Reference Standard, ʟ-Histidine
Catalog # 73767-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    71-00-1
  • Description:
    ʟ-Histidine
  • Storage conditions:
    15...25 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00064315
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    (S)-2-Amino-3-(4-imidazolyl)propionic acid; NSC 137773
  • Melting point:
    282 °C (dec.) (lit.)
  • Molecular weight:
    155,15

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.