Order Entry
Northern Ireland
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, Butyraldehyde-2,4-DNPH
  69630-50MG
 :  Merck
 :  
undefined
Supelco® TraceCERT® Organic Reference Standard, Butyraldehyde-2,4-DNPH
  69630-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    1527-98-6
  • Description:
    Butyraldehyde-2,4-DNPH
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00191327
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    Butyraldehyde-2,4-dinitrophenylhydrazone
  • Melting point:
    117 - 119 °C (lit.)
  • Formula:
    CH₃CH₂CH₂CH=NNHC₆H₃(NO₂)₂
  • Molecular weight:
    252,23

 

 

  • GHS signal: GHS07
 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
 : P305 + P351 + P338