Supelco® Drug and metabolite standards, Trenbolone solution, (CRM)
T-043-1ML
: Merck
:
- Concentration:1,0 mg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:10161-33-8
- Description:Trenbolone solution
- Description:Trenbolone solution
- Pk Info:ampoule of 1 ml