Supelco® Cerilliant® Organic Reference Standard, Prednisone solution
Catalog # P-122-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:53-03-2
- Description:Prednisone solution
- Storage conditions:–20 °C
- Hazard class description:H225 - H302 + H312 + H332 - H319
- Shipping conditions:WICE
- MDL No.:MFCD00003608
- M-Clarity quality level:MQ300
- Melting point:–48 °C
- Molecular weight:358,43
- Density:0,78 g/cm³ at 20 °C
- Flash point:2,0 °C - Closed cup
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338