Order Entry
Northern Ireland
ContactUsLinkComponent
 
Supelco® Cerilliant® Organic Reference Standard, Prednisolone solution
  P-121-1ML
 :  Merck
 :  
undefined
Supelco® Cerilliant® Organic Reference Standard, Prednisolone solution
  P-121-1ML
 :  Merck
 :  

 

  • Concentration:
    100 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Certified reference material, Cerilliant®
  • CAS No.:
    50-24-8
  • Description:
    Prednisolone solution
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319
  • Shipping conditions:
    DICE
  • MDL No.:
    MFCD00003649
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    360,44
  • Flash point:
    2,0 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

 

 

  • GHS signal: GHS02, GHS07
 : P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338