Order Entry
Northern Ireland
ContactUsLinkComponent
 
Supelco® Drug and metabolite standards, paroxetine-D6 maleate solution, (CRM)
  P-915-1ML
 :  Merck
 :  
undefined
Supelco® Drug and metabolite standards, paroxetine-D6 maleate solution, (CRM)
  P-915-1ML
 :  Merck
 :  

 

  • Concentration:
    100 µg/ml in methanol (as free base)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    1435728-64-5
  • Description:
    Paroxetine-D6 maleate solution
  • Description:
    Paroxetine-D6 maleate solution
  • Pk Info:
    ampoule of 1 ml