Order Entry
Northern Ireland
ContactUsLinkComponent
 
Supelco® Drug and metabolite standards, oxcarbazepine solution, (CRM)
  O-025-1ML
 :  Merck
 :  
undefined
Supelco® Drug and metabolite standards, oxcarbazepine solution, (CRM)
  O-025-1ML
 :  Merck
 :  

 

  • Concentration:
    1,0 mg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    28721-07-5
  • Description:
    Oxcarbazepine solution
  • Description:
    Oxcarbazepine solution
  • Pk Info:
    ampoule of 1 ml