Supelco® Drug and metabolite standards, norfluoxetine oxalate solution, (CRM)
Catalog # N-923-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:1,0 mg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:107674-50-0
- Description:Norfluoxetine oxalate solution
- Description:Norfluoxetine oxalate solution
- Pk Info:ampoule of 1 ml